Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
oac-55
Entrez Gene ID
178792
Gene Name
protein-coding
Gene Aliases
CELE_Y39H10A.2
Chromosome Location
Chr.V: 3761139 - 3766315 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330807.1 NP_001317839.1 6 - 7 884 93
NM_071640.2 NP_504041.1 6 - 7 878 93

View Full Product Details