Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-30
Entrez Gene ID
187704
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_R08F11.2
Chromosome Location
Chr.V: 3797928 - 3800478 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071649.2 NP_504050.1 1 - 2 54 63

View Full Product Details