Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp-33C8
Entrez Gene ID
187705
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_R08F11.3
Chromosome Location
Chr.V: 3800782 - 3803542 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071650.7 NP_504051.1 3 - 4 1312 76

View Full Product Details