Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-32
Entrez Gene ID
185911
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_F47C10.6
Chromosome Location
Chr.V: 3841794 - 3848250 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071659.4 NP_504060.2 10 - 11 4332 88

View Full Product Details