Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-65
Entrez Gene ID
187096
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_K07C6.8
Chromosome Location
Chr.V: 3921935 - 3922964 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071685.2 NP_504086.2 2 - 3 421 69

View Full Product Details