Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-34A6
Entrez Gene ID
181848
Gene Name
protein-coding
Gene Aliases
CELE_B0213.11
Chromosome Location
Chr.V: 3958160 - 3961358 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269102.3 NP_001256031.1 4 - 5 1030 118
NM_001330821.3 NP_001317794.1 3 - 4 713 118
NM_001373017.2 NP_001360154.1 4 - 5 964 118

View Full Product Details