Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-83
Entrez Gene ID
191990
Gene Name
Seven TM Receptor
Gene Aliases
CELE_B0213.7
Chromosome Location
Chr.V: 3977264 - 3978811 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071705.1 NP_504106.1 6 - 7 951 127

View Full Product Details