Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-12
Entrez Gene ID
183958
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_D2063.2
Chromosome Location
Chr.V: 4329039 - 4332550 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071795.1 NP_504196.1 8 - 9 1473 121

View Full Product Details