Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
figl-1
Entrez Gene ID
178829
Gene Name
protein-coding
Gene Aliases
CELE_F32D1.1
Chromosome Location
Chr.V: 4341415 - 4345999 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071796.5 NP_504197.1 2 - 3 1017 87

View Full Product Details