Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gnrr-5
Entrez Gene ID
186759
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H22D07.1
Chromosome Location
Chr.V: 4451043 - 4454734 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071827.6 NP_504228.2 3 - 4 801 100

View Full Product Details