Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
soc-1
Entrez Gene ID
178855
Gene Name
Multisubstrate adapter protein soc-1
Gene Aliases
CELE_F41F3.2
Chromosome Location
Chr.V: 4628740 - 4646933 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071849.6 NP_504250.2 9 - 10 2012 65
GenBank mRNA AF419335.1 - 5 - 6 1041 65

View Full Product Details