Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgl-3
Entrez Gene ID
178867
Gene Name
protein-coding
Gene Aliases
CELE_C18G1.4
Chromosome Location
Chr.V: 4755979 - 4758613 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071877.6 NP_504278.1 7 - 8 1690 99
NM_071878.5 NP_504279.1 7 - 8 1700 99
GenBank mRNA AB120729.1 - 7 - 8 1672 99

View Full Product Details