Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-9
Entrez Gene ID
178883
Gene Name
protein-coding
Gene Aliases
CELE_T19H12.1
Chromosome Location
Chr.V: 4896028 - 4897969 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071911.6 NP_504312.1 1 - 2 179 157

View Full Product Details