Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-12
Entrez Gene ID
182573
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C13D9.5
Chromosome Location
Chr.V: 4979947 - 4980973 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071938.1 NP_504339.1 2 - 3 335 129

View Full Product Details