Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srr-8
Entrez Gene ID
182571
Gene Name
Serpentine Receptor, class R
Gene Aliases
CELE_C13D9.3
Chromosome Location
Chr.V: 5001686 - 5003345 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322556.3 NP_001309480.1 4 - 5 803 153

View Full Product Details