Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srr-6
Entrez Gene ID
182569
Gene Name
Serpentine Receptor, class R
Gene Aliases
CELE_C13D9.1
Chromosome Location
Chr.V: 5012803 - 5015219 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071945.3 NP_504346.1 1 - 2 317 140

View Full Product Details