Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srab-2
Entrez Gene ID
182221
Gene Name
Serpentine Receptor, class AB (class A-like)
Gene Aliases
CELE_C04F5.5
Chromosome Location
Chr.V: 5103042 - 5105034 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071966.1 NP_504367.1 5 - 6 836 71

View Full Product Details