Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sid-1
Entrez Gene ID
178900
Gene Name
Systemic RNA interference defective protein 1
Gene Aliases
CELE_C04F5.1
Chromosome Location
Chr.V: 5116087 - 5130059 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071971.9 NP_504372.2 5 - 6 1467 70
GenBank mRNA AF478687.1 - 4 - 5 1292 70

View Full Product Details