Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-39
Entrez Gene ID
182138
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C03A7.6
Chromosome Location
Chr.V: 5156653 - 5157760 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071980.2 NP_504381.2 4 - 5 632 103

View Full Product Details