Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-69
Entrez Gene ID
181872
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_B0238.8
Chromosome Location
Chr.V: 5263951 - 5265681 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072007.6 NP_504408.3 2 - 3 292 112

View Full Product Details