Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cft-1
Entrez Gene ID
178939
Gene Name
Cystic Fibrosis Transmembrane conductance regulator homolog
Gene Aliases
CELE_C18C4.2
Chromosome Location
Chr.V: 5538800 - 5545789 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072062.5 NP_504463.1 9 - 10 1581 103

View Full Product Details