Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpt-2
Entrez Gene ID
178988
Gene Name
Probable 26S proteasome regulatory subunit 4
Gene Aliases
CELE_F29G9.5
Chromosome Location
Chr.V: 6016049 - 6017669 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072157.7 NP_504558.1 3 - 4 1115 110

View Full Product Details