Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-49
Entrez Gene ID
179001
Gene Name
One cut domain family member
Gene Aliases
CELE_F17A9.6
Chromosome Location
Chr.V: 6118322 - 6120206 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072180.4 NP_504581.1 1 - 2 269 90

View Full Product Details