Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rme-1
Entrez Gene ID
179010
Gene Name
protein-coding
Gene Aliases
CELE_W06H8.1
Chromosome Location
Chr.V: 6208840 - 6217664 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029021.4 NP_001024192.1 6 - 7 2025 73
NM_001029023.3 NP_001024194.1 4 - 5 1316 73
NM_001029024.4 NP_001024195.1 4 - 5 1232 73
NM_001029025.6 NP_001024196.1 4 - 5 1359 73
NM_001356657.3 NP_001343865.1 4 - 4 1328 73
NM_001356658.3 NP_001343866.1 4 - 4 1328 73
NM_001356659.3 NP_001343867.1 4 - 4 1210 73
NM_001380664.2 NP_001367910.1 5 - 6 1970 73
NM_001383317.1 4 - 5 1258 73
GenBank mRNA AF357876.1 - 4 - 5 1232 73
AF357878.1 - 4 - 5 1316 73

View Full Product Details