Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rme-1
Entrez Gene ID
179010
Gene Name
protein-coding
Gene Aliases
CELE_W06H8.1
Chromosome Location
Chr.V: 6208840 - 6217664 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029021.4 NP_001024192.1 3 - 4 816 95
NM_001356657.3 NP_001343865.1 1 - 1 119 95
NM_001356658.3 NP_001343866.1 1 - 1 119 95
NM_001380664.2 NP_001367910.1 2 - 3 761 95

View Full Product Details