Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srn-1
Entrez Gene ID
191957
Gene Name
Serpentine Receptor, class N
Gene Aliases
CELE_K12B6.5
Chromosome Location
Chr.V: 6284833 - 6286443 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072206.2 NP_504607.1 5 - 6 780 149

View Full Product Details