Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpt-6
Entrez Gene ID
179035
Gene Name
Carn_acyltransf domain-containing protein
Gene Aliases
CELE_W01A11.5
Chromosome Location
Chr.V: 6490077 - 6493921 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072247.6 NP_504648.1 2 - 3 377 96

View Full Product Details