Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-32
Entrez Gene ID
184705
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F20A1.3
Chromosome Location
Chr.V: 7017359 - 7018851 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072377.2 NP_504778.2 1 - 2 102 102

View Full Product Details