Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-40
Entrez Gene ID
184292
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_F10D2.5
Chromosome Location
Chr.V: 7139475 - 7141964 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072409.7 NP_504810.2 5 - 6 971 98

View Full Product Details