Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-36
Entrez Gene ID
184266
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_F09G2.6
Chromosome Location
Chr.V: 7167009 - 7169281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072419.5 NP_504820.2 7 - 8 1315 130

View Full Product Details