Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
try-8
Entrez Gene ID
179134
Gene Name
Peptidase S1 domain-containing protein
Gene Aliases
CELE_F25E5.10
Chromosome Location
Chr.V: 7452912 - 7454336 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072515.3 NP_504916.1 1 - 2 138 88

View Full Product Details