Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-92
Entrez Gene ID
191879
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_F25E5.13
Chromosome Location
Chr.V: 7459604 - 7460998 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072517.2 NP_504918.1 1 - 2 215 143

View Full Product Details