Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-31
Entrez Gene ID
179136
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C54F6.10
Chromosome Location
Chr.V: 7506869 - 7508666 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072531.2 NP_504932.1 2 - 3 1160 101

View Full Product Details