Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-26
Entrez Gene ID
183662
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_C50E3.10
Chromosome Location
Chr.V: 7600609 - 7601543 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072562.1 NP_504963.1 2 - 3 309 158

View Full Product Details