Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-45
Entrez Gene ID
184130
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_F07C4.2
Chromosome Location
Chr.V: 7638101 - 7638657 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072576.2 NP_504977.1 1 - 2 348 73

View Full Product Details