Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-32A1
Entrez Gene ID
179156
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C26F1.2
Chromosome Location
Chr.V: 7781050 - 7784595 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072608.11 NP_505009.4 3 - 4 430 147

View Full Product Details