Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncx-10
Entrez Gene ID
190821
Gene Name
protein-coding
Gene Aliases
CELE_Y97E10B.7
Chromosome Location
Chr.V: 7930475 - 7933098 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269165.1 NP_001256094.1 2 - 3 432 83
NM_001373058.1 NP_001360010.1 3 - 4 387 83

View Full Product Details