Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-3
Entrez Gene ID
190823
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y97E10B.9
Chromosome Location
Chr.V: 7936144 - 7937252 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072640.1 NP_505041.1 1 - 2 133 144

View Full Product Details