Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpb-9
Entrez Gene ID
179178
Gene Name
TFIIS-type domain-containing protein
Gene Aliases
CELE_Y97E10AR.5
Chromosome Location
Chr.V: 8010812 - 8012345 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072661.6 NP_505062.1 1 - 2 182 69

View Full Product Details