Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asp-6
Entrez Gene ID
179209
Gene Name
Aspartic protease 6
Gene Aliases
CELE_F21F8.7
Chromosome Location
Chr.V: 8267511 - 8268944 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072732.9 NP_505133.1 2 - 3 450 71

View Full Product Details