Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vha-18
Entrez Gene ID
179223
Gene Name
Probable V-type proton ATPase subunit H 1
Gene Aliases
CELE_F52E1.10
Chromosome Location
Chr.V: 8411775 - 8413513 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072770.6 NP_505171.1 2 - 3 272 70

View Full Product Details