Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-40
Entrez Gene ID
179279
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_R03H4.1
Chromosome Location
Chr.V: 9016779 - 9019359 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072932.4 NP_505333.1 4 - 5 649 118

View Full Product Details