Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-50
Entrez Gene ID
179284
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_T27E4.6
Chromosome Location
Chr.V: 9069857 - 9073540 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322580.2 NP_001309487.1 11 - 12 1658 82
NR_136394.1 10 - 11 1459 82

View Full Product Details