Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-31
Entrez Gene ID
179291
Gene Name
protein-coding
Gene Aliases
CELE_F21C10.8
Chromosome Location
Chr.V: 9109581 - 9111013 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028658.4 NP_001023829.1 1 - 2 91 80

View Full Product Details