Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgef-1
Entrez Gene ID
179344
Gene Name
protein-coding
Gene Aliases
CELE_F25B3.3
Chromosome Location
Chr.V: 9556027 - 9561674 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276913.2 NP_001263842.1 7 - 8 980 87
NM_073068.4 NP_505469.3 7 - 8 977 87

View Full Product Details