Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hcp-2
Entrez Gene ID
179353
Gene Name
HoloCentric chromosome binding Protein
Gene Aliases
CELE_T06E4.1
Chromosome Location
Chr.V: 9645746 - 9650211 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073088.4 NP_505489.1 1 - 2 194 63

View Full Product Details