Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
her-1
Entrez Gene ID
179360
Gene Name
protein-coding
Gene Aliases
CELE_ZK287.8
Chromosome Location
Chr.V: 9697015 - 9702035 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029139.3 NP_001024310.1 3 - 4 535 110
NM_001029140.5 NP_001024311.2 1 - 2 167 110

View Full Product Details