Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-7
Entrez Gene ID
179375
Gene Name
C2H2-type domain-containing protein
Gene Aliases
CELE_F46B6.7
Chromosome Location
Chr.V: 9786949 - 9789701 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073125.6 NP_505526.1 1 - 2 179 79

View Full Product Details