Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ruvb-1
Entrez Gene ID
179388
Gene Name
RuvB-like 1
Gene Aliases
CELE_C27H6.2
Chromosome Location
Chr.V: 9978791 - 9980802 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073166.5 NP_505567.2 1 - 2 169 94

View Full Product Details