Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lea-1
Entrez Gene ID
3564838
Gene Name
protein-coding
Gene Aliases
CELE_K08H10.1
Chromosome Location
Chr.V: 10011923 - 10028168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269231.3 NP_001256160.1 3 - 4 364 70
NM_001269232.3 NP_001256161.1 3 - 4 364 70
NM_001269233.4 NP_001256162.1 3 - 4 364 70
NM_001269234.3 NP_001256163.1 2 - 3 316 70
NM_001269235.3 NP_001256164.1 2 - 3 316 70
NM_001269236.3 NP_001256165.1 2 - 3 316 70
NM_001269240.3 NP_001256169.1 1 - 2 100 70
NM_001269241.4 NP_001256170.1 1 - 2 100 70
NM_001269242.2 NP_001256171.1 1 - 2 118 70
NM_001373085.2 NP_001360513.1 3 - 3 364 70
NM_001373086.2 NP_001360514.1 3 - 3 364 70
NM_001373087.2 NP_001360515.1 2 - 2 316 70
NM_001373088.2 NP_001360516.1 2 - 3 316 70
NM_001373089.2 NP_001360517.1 1 - 1 100 70
NM_001373090.2 NP_001360518.1 1 - 1 100 70
GenBank mRNA AF016513.1 - 1 - 2 132 70

View Full Product Details